Learn More
Electron Microscopy Sciences Critical Point Dryer 4 Specimen Holder
SDP
Catalog No. 5028813
Encompass_Preferred
Machined from solid steel and brass/nickel plated. Incorporates a stainless steel mesh top and bottom to allow for fluid exchange. One section which holds multiples of 4 or large samples, cavities measure: 4-9mm(D)x16mm(H), overall: 24mm(D)x28mm(H).
Catalog No. 50-288-13 Supplier Electron Microscopy Sciences Supplier No. 7019404
Machined from solid steel and brass/nickel plated. Incorporates a stainless steel mesh top and bottom to allow for fluid exchange. One section which holds multiples of 4 or large samples, cavities measure: 4-9mm(D)x16mm(H), overall: 24mm(D)x28mm(H).