Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Sulfocalix[4]Arene

Catalog No. 55523250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-523-250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 55-523-250MG Supplier Sigma Aldrich Supplier No. 55523250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Sulfocalix[4]arene can be used as a reactant to form an inclusion complex with nedaplatin to increase its solubility, stability, and bioavailability for efficient drug delivery. It can also be used to prepare ruthenium nanoparticles embedded materials, which are used as efficient catalysts for the reduction of toxic 'sabrilliant yellow'sa azo dye.

Specifications

CAS 1112-39-6
Density 0.88 g/mL at 25°C
Boiling Point 81.4°C
Molecular Formula C4H12O2Si
Linear Formula Si(OCH3)2(CH3)2
Quantity 250 mg
MDL Number MFCD00025691
Synonym 25,26,27,28-Tetrahydroxycalix[4]arene-5,11,17,23-tetrasulfonic acid; Calix[4]arene-4-sulfonic acid
Molecular Weight (g/mol) 120.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.