Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Sulfothiacalix[4]arene Sodium Salt 98.0+%, TCI America™
SDP

Catalog No. p-7135902 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04771G 1 g
S04775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04771G Supplier TCI America Supplier No. S04771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 211561-04-5
Molecular Formula C24H16O16S8
MDL Number MFCD06797166
Synonym Tetrasodium Tetrathiacalix[4]arenetetrasulfonate, Thiacalix[4]arenetetrasulfonic Acid Tetrasodium Salt

Specifications

Melting Point 390°C
Color White
Quantity 1 g
Formula Weight 904.78
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Sulfothiacalix[4]arene Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.