Learn More
Crescent Chemical Co Inc 4-T-OCTYLPHENOL SOLN 1ML
SDP
Supplier:  Crescent Chemical Co Inc CT3762D.10
Encompass_Preferred
4-t-Octylphenol Single Component Calibration Standard in Dichloromethane at 1000 ug/mL 1mL
Catalog No. 50-694-95