Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(tert-Butoxycarbonyl)amino]phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193958 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43351G 1 g
B4335-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43351G Supplier TCI America Supplier No. B43351G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 4-[(tert-Butoxycarbonyl)amino]phenylboronic Acid (contains varying amounts of Anhydride)
CAS 380430-49-9
Melting Point 204°C
Color White
Molecular Formula C11H16BNO4
Quantity 1 g
MDL Number MFCD02093054
Synonym 4-boc-aminophenyl boronic acid, 4-n-boc-amino phenylboronic acid, 4-tert-butoxycarbonyl amino phenyl boronic acid, 4-tert-butoxycarbonylamino phenylboronic acid, 4-tert-butoxycarbonyl aminophenylboronic acid, 4-boc-amino benzeneboronic acid, 4-aminobenzeneboronic acid, n-boc protected, 4-tert-butoxycarbonyl amino phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 4-tert-butoxycarbonylamino phenyl boronic acid
InChI Key UBVOLHQIEQVXGM-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)B(O)O
IUPAC Name (4-{[(tert-butoxy)carbonyl]amino}phenyl)boronic acid
Molecular Weight (g/mol) 237.06
PubChem CID 3613184
Formula Weight 237.06
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.