Learn More
Medchemexpress LLC (4-(tert-Butoxycarbonyl)phenyl)boronic acid | 850568-54-6 | 99.6% | 222.05 | 10 G
SDP
Catalog No. 5000453550
Encompass_Preferred
(4-(tert-Butoxycarbonyl)phenyl)boronic acid is a biochemical reagent used in life science research. It can serve as a biological material or organic compound, enabling various experimental applications. This compound is for research use only and not intended for patient use.
  • Biochemical reagent for life science research
  • Mediates ortho specific hydroxylalkylation of phenols by aldehydes
  • Utilized for fluorescent detection of saccharides by binding diol functional groups
  • Effective catalyst for amidation and esterification of carboxylic acids
Catalog No. 50-004-53550 Supplier Medchemexpress LLC Supplier No. HYW01240810G
(4-(tert-Butoxycarbonyl)phenyl)boronic acid is a biochemical reagent used in life science research. It can serve as a biological material or organic compound, enabling various experimental applications. This compound is for research use only and not intended for patient use.
  • Biochemical reagent for life science research
  • Mediates ortho specific hydroxylalkylation of phenols by aldehydes
  • Utilized for fluorescent detection of saccharides by binding diol functional groups
  • Effective catalyst for amidation and esterification of carboxylic acids