Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(tert-Butoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B47725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4772-5G 5 g
B4772200MG 200 mg
B47721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B4772-5G Supplier TCI America Supplier No. B47725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 850568-54-6
Molecular Formula C11H15BO4
MDL Number MFCD03411946
InChI Key QMVMDYSTJSUDKC-UHFFFAOYSA-N
PubChem CID 2773301

Specifications

Melting Point 106°C
Color White
Quantity 5 g
Synonym 4-tert-butoxycarbonyl phenylboronic acid, 4-tert-butoxycarbonyl phenyl boronic acid, 4-t-butoxycarbonyl phenylboronic acid, tert-butyl 4-boronobenzoate, 4-t-butoxycarbonylphenylboronic acid
SMILES CC(C)(C)OC(=O)C1=CC=C(C=C1)B(O)O
IUPAC Name {4-[(tert-butoxy)carbonyl]phenyl}boronic acid
Molecular Weight (g/mol) 222.05
Formula Weight 222.05
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.