Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. B32485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32485G 5 g
B324825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32485G Supplier TCI America Supplier No. B32485G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 130309-46-5
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
InChI Key KXMRDHPZQHAXML-UHFFFAOYSA-N
Synonym trans-4-boc-amino cyclohexanecarboxylic acid, cis-4-boc-amino cyclohexanecarboxylic acid, 4-boc-amino cyclohexanecarboxylic acid, cis-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-4-aminocyclohexanecarboxylic acid, boc-1,4-trans-achc-oh, cis-4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonylamino cyclohexanecarboxylic acid
PubChem CID 2755996
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 243.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.