Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(tert-Butoxycarbonylamino)cyclohexanone 98.0+%, TCI America™
SDP

Catalog No. B38561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38561G 1 g
B38565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38561G Supplier TCI America Supplier No. B38561G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 179321-49-4
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.28
MDL Number MFCD00798168
InChI Key WYVFPGFWUKBXPZ-UHFFFAOYSA-N
Synonym 4-(Boc-amino)cyclohexanone
PubChem CID 1512535
IUPAC Name tert-butyl N-(4-oxocyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(=O)CC1

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 1 g
Formula Weight 213.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(tert-Butoxycarbonylamino)cyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.