Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(tert-Butoxycarbonylamino)methyl]pyridine-2-carboxylic Acid 96.0+%, TCI America™
SDP

Catalog No. B21241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21241G 1 g
B2124100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21241G Supplier TCI America Supplier No. B21241G
Add to Cart
Add to Cart

[Reagent for application of the exciton chirality method]

Chemical Identifiers

CAS 260970-57-8
Molecular Formula C12H16N2O4
Molecular Weight (g/mol) 252.27
MDL Number MFCD02093452
InChI Key BSNYTBOSHJFWQM-UHFFFAOYSA-N
Synonym 4-(Boc-aminomethyl)pyridine-2-carboxylic Acid

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 252.27
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(tert-Butoxycarbonylamino)methyl]pyridine-2-carboxylic Acid [Reagent for application of the excit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.