Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride, Thermo Scientific Chemicals

Catalog No. AC464350010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464350010 1 g
AC464350050 5 g
AC464350250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC464350010 Supplier Thermo Scientific Chemicals Supplier No. 464350010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947725-04-4
Molecular Formula C12H17F3S
Molecular Weight (g/mol) 250.32
MDL Number MFCD11858620
InChI Key VRTQPEYVMHATOA-UHFFFAOYSA-N
IUPAC Name 5-tert-butyl-1,3-dimethyl-2-(trifluoro-λ⁴-sulfanyl)benzene
SMILES CC1=CC(=CC(C)=C1S(F)(F)F)C(C)(C)C

Specifications

Alpha Vector BUTYLDIMETHYLPHENYLSULFURTRIFLUORIDE
Melting Point 62°C to 68°C
Color Red to White
Boiling Point 92°C to 93°C
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 1 g
UN Number 2923
Sensitivity Moisture sensitive
Solubility Information insoluble aq. soln., soluble in hexane, acetonitrile, dichloromethane
Formula Weight 250.32
Physical Form Crystals or Powder
Chemical Name or Material 4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride
Show More Show Less
Hazard Category GHS05: Corrosive
Hazard Statement GHS06: Toxic
ShelfLife 5 years
storageNote1 normal conditions

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.