Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-tert-butyl-alpha-methyl hydrocinnamic aldehyde | 80-54-6 | MFCD00047655 | 99.2% | 204.31 g/mol | C14H20O | 25 G
SDP

Catalog No. 5000303440
Encompass_Preferred

Lilial (4-tert-butyl-alpha-methyl hydrocinnamic aldehyde) is a synthetic fragrance ingredient provided as a research-grade chemical for laboratory use. It is supplied as a liquid standard with analytical documentation for quality verification and storage guidance.

  • High purity (≈99.2%).
  • Liquid appearance, colorless to light yellow.
  • Molecular formula C14H20O and molecular weight 204.31 g/mol.
  • Available in laboratory pack sizes, including 25 g.
  • Analytical documentation provided: COA, SDS, HNMR, GC.
  • Storage recommendations for stability: in solvent -80°C (6 months) or -20°C (1 month) under nitrogen.
  • Suitable for fragrance formulation research and in vitro toxicology assays.

Catalog No. 50-003-03440 Supplier Medchemexpress LLC Supplier No. 25GHYW014684
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lilial (4-tert-butyl-alpha-methyl hydrocinnamic aldehyde) is a synthetic fragrance ingredient provided as a research-grade chemical for laboratory use. It is supplied as a liquid standard with analytical documentation for quality verification and storage guidance.

  • High purity (≈99.2%).
  • Liquid appearance, colorless to light yellow.
  • Molecular formula C14H20O and molecular weight 204.31 g/mol.
  • Available in laboratory pack sizes, including 25 g.
  • Analytical documentation provided: COA, SDS, HNMR, GC.
  • Storage recommendations for stability: in solvent -80°C (6 months) or -20°C (1 month) under nitrogen.
  • Suitable for fragrance formulation research and in vitro toxicology assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.