Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butyl L-Aspartate 98.0+%, TCI America™
SDP

Catalog No. B39191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39191G 1 g
B39195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39191G Supplier TCI America Supplier No. B39191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3057-74-7
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.211
MDL Number MFCD00038577
InChI Key MXWMFBYWXMXRPD-YFKPBYRVSA-N
Synonym h-asp otbu-oh, l-aspartic acid 4-tert-butyl ester, h-asp obut-oh, 4-tert-butyl hydrogen l-aspartate, 2s-2-amino-4-tert-butoxy-4-oxobutanoic acid, s-2-amino-4-tert-butoxy-4-oxobutanoic acid, aspartic acid, 4-tert-butyl ester, asp but, asp obut, asp otbu oh
PubChem CID 7010570
IUPAC Name (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CC(C(=O)O)N

Specifications

Melting Point 189°C
Color White
Quantity 1 g
Formula Weight 189.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butyl L-Aspartate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.