Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylanisole 98.0+%, TCI America™
SDP

Catalog No. B0961500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0961500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0961500ML Supplier TCI America Supplier No. B0961500ML
Add to Cart
Add to Cart

Specifications

CAS 5396-38-3
Color Yellow
Boiling Point 222°C
Molecular Formula C11H16O
Quantity 500 mL
MDL Number MFCD00026309
Synonym 4-tert-butylanisole, 1-tert-butyl-4-methoxybenzene, p-tert-butylanisole, anisole, p-tert-butyl, benzene, 1-1,1-dimethylethyl-4-methoxy, p-methoxy-tert-butylbenzene, 4-tert-butylphenyl methyl ether, 1-tert-butyl-4-methoxy-benzene, 4-t-butylanisole, 4-tertbutylanisole
InChI Key MCUPBIBNSTXCPQ-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC=C(C=C1)OC
IUPAC Name 1-tert-butyl-4-methoxybenzene
Molecular Weight (g/mol) 164.248
PubChem CID 94750
Formula Weight 164.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylanisole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.