Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-tert-Butylbenzenethiol

Catalog No. 59365625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-365-625G 25 g
59-365-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 59-365-625G Supplier Sigma Aldrich Supplier No. 59365625G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-tert-Butylbenzenethiol can be used to synthesize Pd-thiolate complex by coordinating with Pd2+.The reduction of this complex forms ultrasmall palladium nanoclusters.

Specifications

CAS 2396-68-1
Density 0.964 g/mL (at 25°C (literature))
Boiling Point 238°C (lit.)
Molecular Formula C10H14S
Refractive Index n20/D 1.5480 (literature)
Linear Formula (CH3)3 CC6H4SH
MDL Number MFCD00022067
Quantity 25 g
Synonym 4-tert-Butylthiophenol
Molecular Weight (g/mol) 166.28
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.