Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylcalix[5]arene 98.0+%, TCI America™
SDP

Catalog No. B2221100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2221100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2221100MG Supplier TCI America Supplier No. B2221100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81475-22-1
Molecular Formula C55H70O5
Molecular Weight (g/mol) 811.16
MDL Number MFCD00210028
InChI Key HTJNUHSOASZVHV-UHFFFAOYSA-N
Synonym 4-tert-butylcalix 5 arene, p-tert-butylcalix 5 arene, penta-tert-butyl pentahydroxy calix 5 arene
PubChem CID 2725100
SMILES CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=C(C(=CC(=C5)C(C)(C)C)CC6=C(C(=CC(=C6)C(C)(C)C)C2)O)O)C(C)(C)C)C(C)(C)C)O

Specifications

Color White
Quantity 100 mg
Formula Weight 811.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylcalix[5]arene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.