Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(tert-Butyldimethylsilyl)oxy]benzaldehyde, 97%, Thermo Scientific™

Catalog No. AC428170010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC428170010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428170010 Supplier Thermo Scientific Chemicals Supplier No. 428170010

Chemical Identifiers

CAS 120743-99-9
Molecular Formula C13H20O2Si
Molecular Weight (g/mol) 236.39
MDL Number MFCD05865174
InChI Key XACWSBWCLJXKGI-UHFFFAOYSA-N
Synonym 4-t-butyldimethylsilyloxy benzaldehyde, 4-tert-butyldimethylsilyl oxy benzaldehyde, 4-tert-butyldimethylsilyloxy benzaldehyde, 4-t-butyldimethylsilyl oxy benzaldehyde, 4-tert-butyl dimethyl silyl oxy benzaldehyde, 4-tert-butyldimethylsiloxy benzaldehyde, acmc-209a8a
PubChem CID 10879133
IUPAC Name 4-[tert-butyl(dimethyl)silyl]oxybenzaldehyde
SMILES CC(C)(C)[Si](C)(C)OC1=CC=C(C=O)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 0.9570g/mL
Boiling Point 224°C to 226°C
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 0.957
Formula Weight 236.39
Percent Purity 97%
Chemical Name or Material 4-[(tert-Butyldimethylsilyl)oxy]benzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.