Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(tert-Butyldimethylsilyloxy)-1-butyne 97.0+%, TCI America™
SDP

Catalog No. B50425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50425ML 5 mL
B50421ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50425ML Supplier TCI America Supplier No. B50425ML

Chemical Identifiers

CAS 78592-82-2
Molecular Formula C10H20OSi
Molecular Weight (g/mol) 184.354
MDL Number MFCD03427046
InChI Key NUZBJLXXTAOBPH-UHFFFAOYSA-N
Synonym (But-3-yn-1-yloxy)(tert-butyl)dimethylsilane
PubChem CID 4304567
IUPAC Name tert-butyl-but-3-ynoxy-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCCC#C

Specifications

Color Yellow
Boiling Point 83°C
Quantity 5 mL
UN Number 1993
Formula Weight 184.35
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(tert-Butyldimethylsilyloxy)-1-butyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.