Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylphenol, 99%

Catalog No. aaa1587136
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1587136 500 g
AAA158710E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1587136 Supplier Thermo Scientific Chemicals Supplier No. A1587136
Only null left
Add to Cart

CAS: 98-54-4 | C10H14O | 150.221 g/mol

4-tert-butylphenol on condensation with formaldehyde gives calix[5]arene which is used in enzyme mimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-butylphenol on condensation with formaldehyde gives calix[5]arene which is used in enzyme mimetics.

Solubility
Soluble in water. [8.7 g/L (20°C)]

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-54-4
Molecular Formula C10H14O
MDL Number MFCD00002367
InChI Key QHPQWRBYOIRBIT-UHFFFAOYSA-N
Synonym 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
PubChem CID 7393
ChEBI CHEBI:34444
IUPAC Name 4-tert-butylphenol

Specifications

Melting Point 97°C to 101°C
Density 0.908
Boiling Point 236°C to 238°C
Flash Point 113°C (235°F)
Assay Percent Range 99%
Quantity 500 g
UN Number UN3077
Beilstein 1817334
Merck Index 14,1585
Solubility Information Soluble in water. [8.7g/L (20°C)]
SMILES CC(C)(C)C1=CC=C(C=C1)O
Molecular Weight (g/mol) 150.221
Formula Weight 150.22
Percent Purity 99%
Chemical Name or Material 4-tert-Butylphenol
Show More Show Less
Hazard Category H315-H318-H361f
Hazard Statement GHS H Statement
H318-H317-H335-H315
Precautionary Statement P201-P202-P264b-P281-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-679-0
RTECSNumber SJ8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.