Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-tert-Butylphenylacetylene

Catalog No. 59300125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-300-125G 25 g
59-300-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 59-300-125G Supplier Sigma Aldrich Supplier No. 59300125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-tert-butylphenylacetylene can be synthesized from 4-tert-butylstyrene.4-tert-butylphenylacetylene can be used in the synthesis of the following:
  • 1,6-bis(4-tert-butylphenyl)hexa-1,5-diyne-3,4-dione
  • 4,4',5,5'-tetra(4-tert-butylphenylethynyl)dibenzo-24-crown-8-ether via sonogashira coupling reaction with 4,4',5,5'-tetraiododibenzo-24-crown-8-ether
  • 1-(4-tert-butylphenyl)-2-(4-tert-butyldimethylsiloxyphenyl)acetylene via palladium catalyzed coupling with 4-(tert-butyldimethylsiloxy)iodobenzene

Specifications

CAS 772-38-3
Density 0.877 g/mL (at 25°C (literature))
Boiling Point 70°C (2 mmHg)
Molecular Formula C12H14
Linear Formula (CH3)3C6H4C///CH
MDL Number MFCD00190197
Quantity 25 g
Molecular Weight (g/mol) 158.24
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.