Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-tert-Butylpyridine

Catalog No. 14237925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-237-925G 25 g
14-237-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-237-925G Supplier Sigma Aldrich Supplier No. 14237925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-tert-Butylpyridine is specific additive of redox electrolyte in dye sensitized solar cells and dye-sensitized TiO2 solar cells.4-tert-Butylpyridine was used in composition of electrolyte for dye-sensitized solar cell.

Specifications

CAS 3978-81-2
Density 0.923 g/mL (at 25°C (literature))
Boiling Point 196°C to 197°C (lit.)
Molecular Formula C9H13N
Refractive Index n20/D 1.495 (literature)
Linear Formula C9H13N
MDL Number MFCD00006435
Quantity 25 g
Molecular Weight (g/mol) 135.21
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.