Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylpyridine 96.0+%, TCI America™
SDP

Catalog No. B038825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B038825G 25 g
B03885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B038825G Supplier TCI America Supplier No. B038825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3978-81-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00006435
InChI Key YSHMQTRICHYLGF-UHFFFAOYSA-N
Synonym 4-tert-butyl pyridine, pyridine, 4-1,1-dimethylethyl, 4-t-butylpyridine, pyridine, 4-tert-butyl, p-tert-butyl pyridine, unii-il8v02163i, 4-tert-butyl-pyridine, 4-t-butyl pyridine, 4-tert.butylpyridine
PubChem CID 19878
IUPAC Name 4-tert-butylpyridine
SMILES CC(C)(C)C1=CC=NC=C1

Specifications

Melting Point -40°C
Color Yellow
Boiling Point 197°C
Quantity 25 g
Formula Weight 135.21
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.