Learn More
Medchemexpress LLC 4-tert-butyltoluene | 98-51-1 | MFCD00008837 | 95.0% | 148.24 g/mol | C11H16 | 25 G
SDP
Catalog No. 5000209314
Encompass_Preferred
4-tert-Butyltoluene (p-tert-Butyl toluene) is an aromatic hydrocarbon used as a chemical and pharmaceutical synthesis intermediate for research applications. It is typically supplied at 95.0% purity, has molecular formula C11H16, and a molecular weight of approximately 148.24 g/mol.
  • Aromatic hydrocarbon used as a synthesis intermediate
  • Typical purity 95.0%
  • CAS number 98-51-1 for unambiguous identification
  • Supplied in laboratory-scale quantities, for example 25 G
  • Intended for research use only
Catalog No. 50-002-09314 Supplier Medchemexpress LLC Supplier No. 25GHYW040301
Add to Cart
Add to Cart
4-tert-Butyltoluene (p-tert-Butyl toluene) is an aromatic hydrocarbon used as a chemical and pharmaceutical synthesis intermediate for research applications. It is typically supplied at 95.0% purity, has molecular formula C11H16, and a molecular weight of approximately 148.24 g/mol.
  • Aromatic hydrocarbon used as a synthesis intermediate
  • Typical purity 95.0%
  • CAS number 98-51-1 for unambiguous identification
  • Supplied in laboratory-scale quantities, for example 25 G
  • Intended for research use only