Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Octylphenol, MilliporeSigma™ Supelco™

Catalog No. 111020252 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0252 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0252 Supplier MilliporeSigma Supelco Supplier No. 442858
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 140-66-9
Physical Form Neat
Linear Formula (CH3)3 CCH2 C(CH3)2C6H4 OH
MDL Number MFCD00002368
Melting Point 79°C to 82°C
Boiling Point 175°C (30 mmHg)
Synonym 4-(1,1,3,3-Tetramethylbutyl)phenol
Type Organic Pollutant Standard
Quantity 500 mg
Formula Weight 206.32
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.