Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Octylphenol monoethoxylate Solution, 10 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111018592 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8592 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8592 Supplier MilliporeSigma Supelco Supplier No. 328821ML
Add to Cart
Add to Cart

Sigma-Aldrich offers 4-tert-octylphenol monoethoxylate solution for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyzes.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 2315-67-5
Flash Point -17°C
Linear Formula C16H26O2
MDL Number MFCD00214058
Synonym 2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethanol
Type Analytical Standard
Quantity 1 mL
Formula Weight 250.38
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 10 μg/mL in acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.