Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Octylphenol Solution, 1,000 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111018616 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8616 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8616 Supplier MilliporeSigma Supelco Supplier No. 324671ML
Add to Cart
Add to Cart

4-tert-Octylphenols are classified under the alkylphenol family of compounds.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 140-66-9
Flash Point -17.0°C - Closed Cup
Linear Formula C14H22 O
MDL Number MFCD00002368
Synonym 4-(1,1,3,3-Tetramethylbutyl)phenol solution
Type Analytical Standard
Quantity 1 mL
Formula Weight 206.32
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 1,000 μg/mL in acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.