Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Octylresorcinol 99.0+%, TCI America™
SDP

Catalog No. O004825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O004825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O004825G Supplier TCI America Supplier No. O004825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28122-52-3
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00045770
InChI Key YQOJNENEFSZINP-UHFFFAOYSA-N
Synonym 4-tert-octylresorcinol, 4-1,1,3,3-tetramethylbutyl benzene-1,3-diol, p-tert-octylresorcinol, 4-2,4,4-trimethylpentan-2-yl benzene-1,3-diol, 4-t-octyl resorcinol, acmc-209h0z, 4-1,1,3,3-tetramethylbutyl resorcinol, 1,3-benzenediol,4-1,1,3,3-tetramethylbutyl, 4-1,1,3,3-tetramethylbutyl-1,3-benzenediol, 1,3-benzenediol, 4-1,1,3,3-tetramethylbutyl
PubChem CID 220749
IUPAC Name 4-(2,4,4-trimethylpentan-2-yl)benzene-1,3-diol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(O)C=C1O

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 25 g
Formula Weight 222.33
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Octylresorcinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.