Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Pentylcyclohexanone, 97%

Catalog No. AAA1283230
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1283230 250 g
AAA1283218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1283230 Supplier Thermo Scientific Chemicals Supplier No. A1283230
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 16587-71-6 | C11H20O | 168.28 g/mol

It finds its application as a masking agent and a perfuming agent in cosmetic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a masking agent and a perfuming agent in cosmetic industry.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16587-71-6
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00013798
InChI Key DCSKAMGZSIRJAQ-UHFFFAOYSA-N
Synonym 4-tert-pentyl cyclohexanone, 4-tert-amylcyclohexanone, 4-tert-pentylcyclohexanone, cyclohexanone, 4-1,1-dimethylpropyl, 4-2-methylbutan-2-yl cyclohexan-1-one, 4-1,1-dimethylpropyl cyclohexanone, cyclohexanone, 4-tert-pentyl, orivone, unii-5380bwu79x, 4-tert-pentyl-cyclohexanone
PubChem CID 27941
IUPAC Name 4-(2-methylbutan-2-yl)cyclohexan-1-one
SMILES CCC(C)(C)C1CCC(=O)CC1

Specifications

Density 0.92
Boiling Point 124°C to 125°C (16 mmHg)
Refractive Index 1.469
Quantity 250 g
Beilstein 2240475
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 168.28
Percent Purity 97%
Chemical Name or Material 4-tert-Pentylcyclohexanone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 240-642-0
RTECSNumber GW1960000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.