Learn More
Medchemexpress LLC 4-thiazoleacetic acid, 2-(4-chlorophenyl)- | 17969-20-9 | 99.5% | 253.70 g/mol | C11H8ClNO2S | 25 MG
SDP
Catalog No. 5000249144
Encompass_Preferred
Fenclozic acid is an orally active analgesic, antipyretic, and anti-inflammatory small molecule (CAS 17969-20-9) used in pharmacological research to study pain and inflammatory pathways. It is provided as a high-purity research chemical with available analytical documentation for experimental use. Molecular formula C11H8ClNO2S; molecular weight 253.70 g/mol.
  • Orally active analgesic, antipyretic, and anti-inflammatory properties.
  • High purity (~99.5%) appropriate for research applications.
  • Available in small pack sizes for screening and dosing.
  • Supplied with datasheet and certificate of analysis for quality verification.
  • Recommended storage conditions provided to support stability.
Catalog No. 50-002-49144 Supplier Medchemexpress LLC Supplier No. HY10143225MG
Add to Cart
Add to Cart
Fenclozic acid is an orally active analgesic, antipyretic, and anti-inflammatory small molecule (CAS 17969-20-9) used in pharmacological research to study pain and inflammatory pathways. It is provided as a high-purity research chemical with available analytical documentation for experimental use. Molecular formula C11H8ClNO2S; molecular weight 253.70 g/mol.
  • Orally active analgesic, antipyretic, and anti-inflammatory properties.
  • High purity (~99.5%) appropriate for research applications.
  • Available in small pack sizes for screening and dosing.
  • Supplied with datasheet and certificate of analysis for quality verification.
  • Recommended storage conditions provided to support stability.