Learn More
Medchemexpress LLC 4-Thiazolecarboxamide, N-(2-aminoethyl)-5-(3-fluorophenyl)-, hydrochloride (1:1) | 127917-66-2 | MFCD00900596 | 100.0% | 301.77 | C12H13ClFN3OS | 50 MG
SDP
Catalog No. 5000215015
Encompass_Preferred
Ro 41-1049 hydrochloride is a reversible, selective monoamine oxidase-A (MAO-A) inhibitor supplied as a solid research reagent with high reported purity. It has documented in vitro binding (Kd ≈16.5 nM in human frontal cortex) and shown in vivo effects on dopamine metabolism. Solubility data indicate good solubility in DMSO and water. The product is provided with supporting analytical documents for quality verification.
  • High reported purity suitable for research applications.
  • Selective, reversible inhibition of MAO-A.
  • Documented solubility in common solvents (DMSO, water).
  • Provided with COA, SDS, and datasheet for quality assurance.
  • Available in multiple pack sizes for experimental flexibility.
Catalog No. 50-002-15015 Supplier Medchemexpress LLC Supplier No. HY100027A50MG
Ro 41-1049 hydrochloride is a reversible, selective monoamine oxidase-A (MAO-A) inhibitor supplied as a solid research reagent with high reported purity. It has documented in vitro binding (Kd ≈16.5 nM in human frontal cortex) and shown in vivo effects on dopamine metabolism. Solubility data indicate good solubility in DMSO and water. The product is provided with supporting analytical documents for quality verification.
  • High reported purity suitable for research applications.
  • Selective, reversible inhibition of MAO-A.
  • Documented solubility in common solvents (DMSO, water).
  • Provided with COA, SDS, and datasheet for quality assurance.
  • Available in multiple pack sizes for experimental flexibility.