Learn More
Medchemexpress LLC 4-thiazolecarboxylic acid, 2-[8-[(2-benzothiazolylamino)carbonyl]-3,4-dihydro-2(1h)-isoquino | 1235032-75-3 | 98.9% | 25 MG
SDP
Catalog No. 5000341607
Encompass_Preferred
Bcl-xL antagonist 2 is a potent, selective, and orally active antagonist of BCL-XL with an IC50 of 0.091 μM and a Ki of 65 nM. It promotes the apoptosis of cancer cells.
  • Potent and selective antagonist of BCL-XL.
  • Orally active.
  • Promotes the apoptosis of cancer cells.
  • Potential for research of chronic lymphocytic leukemia (CLL) and non-Hodgkin's lymphoma (NHL).
Catalog No. 50-003-41607 Supplier Medchemexpress LLC Supplier No. 25MGHY12908
Bcl-xL antagonist 2 is a potent, selective, and orally active antagonist of BCL-XL with an IC50 of 0.091 μM and a Ki of 65 nM. It promotes the apoptosis of cancer cells.
  • Potent and selective antagonist of BCL-XL.
  • Orally active.
  • Promotes the apoptosis of cancer cells.
  • Potential for research of chronic lymphocytic leukemia (CLL) and non-Hodgkin's lymphoma (NHL).