Learn More
Medchemexpress LLC 4-Thiazolecarboxylic acid | 3973-08-8 | 99.97% | C4H3NO2S | 10 G
SDP
Catalog No. 5000433814
Encompass_Preferred
4-Thiazolecarboxylic acid (Compound 1e) is an intermediate used in the synthesis of MGL-IN-2. MGL-IN-2 inhibits wild-type MGL enzyme activity with an IC50 of 0.0121 μM. This compound is suitable for research into metabolic diseases, acting as a biochemical assay reagent and drug intermediate.
  • An intermediate compound.
  • Used in the synthesis of MGL-IN-2.
  • Applicable in metabolic disease research.
  • Functions as a biochemical assay reagent.
  • Serves as a drug intermediate.
Catalog No. 50-004-33814 Supplier Medchemexpress LLC Supplier No. HY5932010G
4-Thiazolecarboxylic acid (Compound 1e) is an intermediate used in the synthesis of MGL-IN-2. MGL-IN-2 inhibits wild-type MGL enzyme activity with an IC50 of 0.0121 μM. This compound is suitable for research into metabolic diseases, acting as a biochemical assay reagent and drug intermediate.
  • An intermediate compound.
  • Used in the synthesis of MGL-IN-2.
  • Applicable in metabolic disease research.
  • Functions as a biochemical assay reagent.
  • Serves as a drug intermediate.