Learn More
Medchemexpress LLC 4-Thiazolecarboxylic acid | 3973-08-8 | 99.97% | C4H3NO2S | 25 G
SDP
Catalog No. 5000357615
Encompass_Preferred
4-Thiazolecarboxylic acid is an intermediate chemical compound. It can be utilized in the synthesis of MGL-IN-2, which is known to inhibit wild-type MGL enzyme activity with an IC50 of 0.0121 μM. This compound is also suitable for research applications concerning metabolic diseases.
  • Intermediate chemical compound
  • Used in MGL-IN-2 synthesis
  • Supports research in metabolic diseases
Catalog No. 50-003-57615 Supplier Medchemexpress LLC Supplier No. 25GHY59320
4-Thiazolecarboxylic acid is an intermediate chemical compound. It can be utilized in the synthesis of MGL-IN-2, which is known to inhibit wild-type MGL enzyme activity with an IC50 of 0.0121 μM. This compound is also suitable for research applications concerning metabolic diseases.
  • Intermediate chemical compound
  • Used in MGL-IN-2 synthesis
  • Supports research in metabolic diseases