Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-thien-2-ylphenyl)methanol, Thermo Scientific™

Catalog No. CC12009DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC12009DA 1 g Amber Glass Bottle
CC12009CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC12009DA Supplier Thermo Scientific Chemicals Supplier No. CC12009DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81443-44-9
Molecular Formula C11H10OS
Molecular Weight (g/mol) 190.26
InChI Key KHZSWSHUIYOMSH-UHFFFAOYSA-N
Synonym 4-thien-2-ylphenyl methanol, 4-thiophen-2-yl phenyl methanol, thiophene,2-4-methylphenyl, benzenemethanol, 4-2-thienyl, 4-thien-2-yl-phenyl methanol, pubchem24308, 4-2-thienyl benzyl alcohol, 4-thien-2-yl benzyl alcohol, 4-2-thienyl phenyl methanol
PubChem CID 2795254
IUPAC Name (4-thiophen-2-ylphenyl)methanol
SMILES C1=CSC(=C1)C2=CC=C(C=C2)CO

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.26
Percent Purity 97%
Chemical Name or Material (4-thien-2-ylphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.