Learn More
Medchemexpress LLC 4-thio-2'-deoxyuridine | 5580-20-1 | MFCD00058457 | 98.0% | 244.27 g·mol⁻¹ | C9H12N2O4S | 5 MG
SDP
Catalog No. 5000281217
Encompass_Preferred
4-Thio-2'-deoxyuridine is a thio-substituted deoxyuridine nucleoside analog used as a research reagent in studies of nucleic acid chemistry, photophysics, and biochemical assays. It serves as a probe for investigating nucleic acid structure, reactivity, and photoinduced processes in vitro.
  • Thio-substituted nucleoside analog for nucleic acid research.
  • Molecular formula C9H12N2O4S; molecular weight 244.27 g·mol⁻¹.
  • High purity (≥98%) suitable for biochemical assays.
  • Useful in photochemical and antimetabolite studies.
  • Comes with manufacturer data sheet and safety documentation for laboratory handling.
Catalog No. 50-002-81217 Supplier Medchemexpress LLC Supplier No. HYW25178150MG
4-Thio-2'-deoxyuridine is a thio-substituted deoxyuridine nucleoside analog used as a research reagent in studies of nucleic acid chemistry, photophysics, and biochemical assays. It serves as a probe for investigating nucleic acid structure, reactivity, and photoinduced processes in vitro.
  • Thio-substituted nucleoside analog for nucleic acid research.
  • Molecular formula C9H12N2O4S; molecular weight 244.27 g·mol⁻¹.
  • High purity (≥98%) suitable for biochemical assays.
  • Useful in photochemical and antimetabolite studies.
  • Comes with manufacturer data sheet and safety documentation for laboratory handling.