Learn More
Medchemexpress LLC 4-thio-2'-deoxyuridine | 5580-20-1 | MFCD00058457 | 99.8% | 244.27 g/mol | C9H12N2O4S | 100 MG
SDP
Catalog No. 5000281524
Encompass_Preferred
4-Thio-2'-deoxyuridine is a sulfur-substituted deoxyuridine nucleoside analogue used in biochemical and nucleic acid research, including studies of nucleic acid chemistry and incorporation into oligonucleotides. It has molecular formula C9H12N2O4S, molecular weight 244.27 g/mol, and a reported purity of 99.78%.
  • Sulfur-substituted deoxyuridine analogue useful for nucleic acid studies.
  • High reported purity (99.8%), suitable for analytical and synthetic applications.
  • Available in multiple pack sizes for research-scale work.
  • Provided with certificate of analysis and safety data sheet for quality and handling.
  • Offered in solid form and as 10 mM DMSO solution for convenient use.
Catalog No. 50-002-81524 Supplier Medchemexpress LLC Supplier No. HYW251781100MG
4-Thio-2'-deoxyuridine is a sulfur-substituted deoxyuridine nucleoside analogue used in biochemical and nucleic acid research, including studies of nucleic acid chemistry and incorporation into oligonucleotides. It has molecular formula C9H12N2O4S, molecular weight 244.27 g/mol, and a reported purity of 99.78%.
  • Sulfur-substituted deoxyuridine analogue useful for nucleic acid studies.
  • High reported purity (99.8%), suitable for analytical and synthetic applications.
  • Available in multiple pack sizes for research-scale work.
  • Provided with certificate of analysis and safety data sheet for quality and handling.
  • Offered in solid form and as 10 mM DMSO solution for convenient use.