Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-thio-2'-deoxyuridine | 5580-20-1 | MFCD00058457 | 99.8% | 244.27 g·mol⁻¹ | C9H12N2O4S | 25 MG
SDP

Catalog No. 5000281605
Encompass_Preferred

4-Thio-2'-deoxyuridine is a purine nucleoside analog used in biochemical and cellular research to probe nucleic acid chemistry and antitumor mechanisms. It can inhibit DNA synthesis and promote apoptosis in experimental systems. Supplied as a light yellow solid with high reported purity, the compound is available in small milligram packages and as ready-to-use DMSO solutions for assay development and screening.

  • Purine nucleoside analog for research applications.
  • Molecular formula C9H12N2O4S; MW 244.27 g·mol⁻¹.
  • CAS number 5580-20-1 for unambiguous identification.
  • High purity (reported 99.78%).
  • Solid form; light yellow to yellow appearance.
  • Storage recommendations provided for powder and solvent solutions.
  • Available in small packaging sizes, including 25 mg.
  • Concentration options supplied for solution-based assays (e.g., DMSO stocks).

Catalog No. 50-002-81605 Supplier Medchemexpress LLC Supplier No. HYW25178125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Thio-2'-deoxyuridine is a purine nucleoside analog used in biochemical and cellular research to probe nucleic acid chemistry and antitumor mechanisms. It can inhibit DNA synthesis and promote apoptosis in experimental systems. Supplied as a light yellow solid with high reported purity, the compound is available in small milligram packages and as ready-to-use DMSO solutions for assay development and screening.

  • Purine nucleoside analog for research applications.
  • Molecular formula C9H12N2O4S; MW 244.27 g·mol⁻¹.
  • CAS number 5580-20-1 for unambiguous identification.
  • High purity (reported 99.78%).
  • Solid form; light yellow to yellow appearance.
  • Storage recommendations provided for powder and solvent solutions.
  • Available in small packaging sizes, including 25 mg.
  • Concentration options supplied for solution-based assays (e.g., DMSO stocks).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.