Learn More
Apexbio Technology LLC 4-Thio-dTTP 100ul (100 mM)
SDP
Catalog No. 5000087776
Encompass_Preferred
4-Thio-dTTP is a nucleotide analog structurally similar to thymidine triphosphate (dTTP) It incorporates into DNA strands during enzymatic synthesis modifying nucleic acid interactions and affecting the activities of enzymes involved in DNA replication and repair 4-Thio-dTTP exerts its biological activity through incorporation into DNA enabling the study of DNA polymerase fidelity replication kinetics enzyme-substrate interactions and mechanisms of DNA damage responses Based on these biochemical properties 4-Thio-dTTP holds research potential in applications such as investigating DNA synthesis regulation enzymatic mechanisms and pathways related to cancer and genomic instability
Catalog No. 50-000-87776 Supplier Apexbio Technology LLC Supplier No. B81030.1
4-Thio-dTTP is a nucleotide analog structurally similar to thymidine triphosphate (dTTP) It incorporates into DNA strands during enzymatic synthesis modifying nucleic acid interactions and affecting the activities of enzymes involved in DNA replication and repair 4-Thio-dTTP exerts its biological activity through incorporation into DNA enabling the study of DNA polymerase fidelity replication kinetics enzyme-substrate interactions and mechanisms of DNA damage responses Based on these biochemical properties 4-Thio-dTTP holds research potential in applications such as investigating DNA synthesis regulation enzymatic mechanisms and pathways related to cancer and genomic instability