Learn More
Cayman Chemical 4-Thiouracil, Purity: ≥98%, CAS: 591-28-6, 1 g
SDP
Catalog No. 502968109
Encompass_Preferred
A site-specific, photoactivatable probe used to detect RNA structures and nucleic acid-nucleic acid contacts
Catalog No. 50-296-8109 Supplier Cayman Chemical Supplier No. 169521 G
A site-specific, photoactivatable probe used to detect RNA structures and nucleic acid-nucleic acid contacts