Learn More
Cayman Chemical 4-Thiouracil, Purity: ≥98%, CAS: 591-28-6, 500 mg
SDP
Catalog No. 502965111
Encompass_Preferred
A site-specific, photoactivatable probe used to detect RNA structures and nucleic acid-nucleic acid contacts
Catalog No. 50-296-5111 Supplier Cayman Chemical Supplier No. 16952500 MG
A site-specific, photoactivatable probe used to detect RNA structures and nucleic acid-nucleic acid contacts