Learn More
Medchemexpress LLC 4-Thiouracil | 591-28-6 | 99.98% | 128.15 | 10 G
SDP
Catalog No. 5000452562
Encompass_Preferred
4-Thiouracil is a thionucleobase with cytostatic properties. It can be utilized as biological photoprobes for detecting RNA structures and nucleic acid-nucleic acid contacts. Additionally, it functions as a potent ultraviolet A (UVA) photosensitizer, generating reactive oxygen species (O2). 4-Thiouracil shows promise for research in photocross linking, photodamage, and photodynamic therapy.
  • Thionucleobase with cytostatic properties.
  • Can be used as biological photoprobes to detect RNA structures and nucleic acid-nucleic acid contacts.
  • Acts as a strong ultraviolet A (UVA) photosensitizer, providing a source of the reactive oxygen species of O2.
  • Promising for research of photocross linking, photodamage, as well as photodynamic therapy.
Catalog No. 50-004-52562 Supplier Medchemexpress LLC Supplier No. HYW01041710G
4-Thiouracil is a thionucleobase with cytostatic properties. It can be utilized as biological photoprobes for detecting RNA structures and nucleic acid-nucleic acid contacts. Additionally, it functions as a potent ultraviolet A (UVA) photosensitizer, generating reactive oxygen species (O2). 4-Thiouracil shows promise for research in photocross linking, photodamage, and photodynamic therapy.
  • Thionucleobase with cytostatic properties.
  • Can be used as biological photoprobes to detect RNA structures and nucleic acid-nucleic acid contacts.
  • Acts as a strong ultraviolet A (UVA) photosensitizer, providing a source of the reactive oxygen species of O2.
  • Promising for research of photocross linking, photodamage, as well as photodynamic therapy.