Learn More
Medchemexpress LLC 4-Thiouracil | 591-28-6 | 99.98% | 128.15 | 100 MG
SDP
Catalog No. 5000367052
Encompass_Preferred
4-Thiouracil is a thionucleobase possessing cytostatic properties. It functions as a biological photoprobe for detecting RNA structures and nucleic acid-nucleic acid contacts. Additionally, it serves as a potent ultraviolet A (UVA) photosensitizer, generating reactive oxygen species (O2). 4-Thiouracil holds promise for research in photocrosslinking, photodamage, and photodynamic therapy.
  • Thionucleobase with cytostatic properties.
  • Can be used as biological photoprobes to detect RNA structures and nucleic acid-nucleic acid contacts.
  • Acts as a strong UVA photosensitizer, providing a source of reactive oxygen species (O2).
  • Promising for research in photocrosslinking, photodamage, and photodynamic therapy.
Catalog No. 50-003-67052 Supplier Medchemexpress LLC Supplier No. HYW010417100MG
4-Thiouracil is a thionucleobase possessing cytostatic properties. It functions as a biological photoprobe for detecting RNA structures and nucleic acid-nucleic acid contacts. Additionally, it serves as a potent ultraviolet A (UVA) photosensitizer, generating reactive oxygen species (O2). 4-Thiouracil holds promise for research in photocrosslinking, photodamage, and photodynamic therapy.
  • Thionucleobase with cytostatic properties.
  • Can be used as biological photoprobes to detect RNA structures and nucleic acid-nucleic acid contacts.
  • Acts as a strong UVA photosensitizer, providing a source of reactive oxygen species (O2).
  • Promising for research in photocrosslinking, photodamage, and photodynamic therapy.