Learn More
Medchemexpress LLC 4-Thiouracil | 591-28-6 | 99.98% | 128.15 | 250 MG
SDP
Catalog No. 5000453413
Encompass_Preferred
4-Thiouracil is a thionucleobase with cytostatic properties. It can be utilized as a biological photoprobe to detect RNA structures and nucleic acid-nucleic acid contacts. Additionally, it functions as a strong ultraviolet A (UVA) photosensitizer, serving as a source of reactive oxygen species (O2). 4-Thiouracil shows promise for research in photocrosslinking, photodamage, and photodynamic therapy.
  • Thionucleobase with cytostatic properties
  • Can be used as biological photoprobes to detect RNA structures and nucleic acid-nucleic acid contacts
  • Acts as a strong ultraviolet A (UVA) photosensitizer
  • Provides a source of reactive oxygen species of O2
  • Promising for research of photocrosslinking, photodamage, and photodynamic therapy
Catalog No. 50-004-53413 Supplier Medchemexpress LLC Supplier No. 250MGHYW010417
4-Thiouracil is a thionucleobase with cytostatic properties. It can be utilized as a biological photoprobe to detect RNA structures and nucleic acid-nucleic acid contacts. Additionally, it functions as a strong ultraviolet A (UVA) photosensitizer, serving as a source of reactive oxygen species (O2). 4-Thiouracil shows promise for research in photocrosslinking, photodamage, and photodynamic therapy.
  • Thionucleobase with cytostatic properties
  • Can be used as biological photoprobes to detect RNA structures and nucleic acid-nucleic acid contacts
  • Acts as a strong ultraviolet A (UVA) photosensitizer
  • Provides a source of reactive oxygen species of O2
  • Promising for research of photocrosslinking, photodamage, and photodynamic therapy