Learn More
Cayman Chemical 4-ThIouracIl 5g
SDP
Catalog No. 502966434
Encompass_Preferred
A site-specific, photoactivatable probe used to detect RNA structures and nucleic acid-nucleic acid contacts
Catalog No. 50-296-6434 Supplier Cayman Chemical Supplier No. 169525 G
A site-specific, photoactivatable probe used to detect RNA structures and nucleic acid-nucleic acid contacts