Learn More
Cayman Chemical 4-Thiouridine, Purity: ≥98%, CAS: 13957-31-8, 100 mg
SDP
Catalog No. 502970221
Encompass_Preferred
A photoactivatable ribonucleoside analog that is widely used for RNA analysis, including short-range RNA-RNA crosslinking and nascent RNA labeling
Catalog No. 50-297-0221 Supplier Cayman Chemical Supplier No. 16373100 MG
A photoactivatable ribonucleoside analog that is widely used for RNA analysis, including short-range RNA-RNA crosslinking and nascent RNA labeling