Learn More
Medchemexpress LLC 4-Thiouridine 10mg
SDP
Catalog No. 5000206628
Encompass_Preferred
4 -Thiouridine (1-(4-Thio-beta-D-ribofuranosyl)uracil) is a thio analog of thymidine 4 -Thiouridine can be used to synthesize 4 -thionucleosides with antiviral and anticancer activities 4 -Thiouridine can be used for RNA-sequencing[1][2][3]
Catalog No. 50-002-06628 Supplier Medchemexpress LLC Supplier No. HYW11308110MG
4 -Thiouridine (1-(4-Thio-beta-D-ribofuranosyl)uracil) is a thio analog of thymidine 4 -Thiouridine can be used to synthesize 4 -thionucleosides with antiviral and anticancer activities 4 -Thiouridine can be used for RNA-sequencing[1][2][3]