Learn More
Medchemexpress LLC 4-Thiouridine | 13957-31-8 | 99.98% | 260.27 | 25 MG
SDP
Catalog No. 5000452127
Encompass_Preferred
4-Thiouridine is a ribonucleoside analog, widely used in RNA analysis and (m)RNA labeling. It inhibits rRNA synthesis and causes a nucleolar stress response.
  • Functions as a ribonucleoside analog.
  • Utilized for RNA analysis and (m)RNA labeling.
  • Labels and purifies nascent rRNA at concentrations of 0-10 μM without significantly affecting rRNA synthesis.
  • Strongly inhibits the production and processing of rRNA at concentrations of ≥50 μM.
  • Targets nucleoside antimetabolite/analog.
  • Acts on the cell cycle/DNA damage pathway.
Catalog No. 50-004-52127 Supplier Medchemexpress LLC Supplier No. 25MGHYW011793
4-Thiouridine is a ribonucleoside analog, widely used in RNA analysis and (m)RNA labeling. It inhibits rRNA synthesis and causes a nucleolar stress response.
  • Functions as a ribonucleoside analog.
  • Utilized for RNA analysis and (m)RNA labeling.
  • Labels and purifies nascent rRNA at concentrations of 0-10 μM without significantly affecting rRNA synthesis.
  • Strongly inhibits the production and processing of rRNA at concentrations of ≥50 μM.
  • Targets nucleoside antimetabolite/analog.
  • Acts on the cell cycle/DNA damage pathway.