Learn More
Apexbio Technology LLC 4-Thiouridine(Synonyms: 4-Thio-uridine, 4SU, s4U, 4-thiouracil riboside, 4-thio-ribouridine), 25mg, CAS: 13957-31-8.
SDP
Catalog No. 5000060930
Encompass_Preferred
4-Thiouridine (CAS 13957-31-8) is a thiol-substituted ribonucleoside analog commonly utilized in RNA research for photoactivatable labeling and RNA cross-linking experiments Structurally 4-thiouridine differs from natural uridine solely by the substitution of a sulfur atom at the 4-position of the nucleotide base Due to this close structural similarity it integrates into RNA sequences with minimal disruption preserving typical base-pairing properties and RNA functionality Upon UV irradiation the incorporated thiol group induces covalent short-range RNA RNA cross-links facilitating detailed analysis of RNA interactions turnover rates and dynamics within cellular contexts
Catalog No. 50-000-60930 Supplier Apexbio Technology LLC Supplier No. C372225
4-Thiouridine (CAS 13957-31-8) is a thiol-substituted ribonucleoside analog commonly utilized in RNA research for photoactivatable labeling and RNA cross-linking experiments Structurally 4-thiouridine differs from natural uridine solely by the substitution of a sulfur atom at the 4-position of the nucleotide base Due to this close structural similarity it integrates into RNA sequences with minimal disruption preserving typical base-pairing properties and RNA functionality Upon UV irradiation the incorporated thiol group induces covalent short-range RNA RNA cross-links facilitating detailed analysis of RNA interactions turnover rates and dynamics within cellular contexts