Learn More
TARGETMOL CHEMICALS INC 4-Thiouridine 25MG
SDP
Catalog No. 502878729
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 4-Thiouridine (4-SU) is a photoactivatable ribonucleoside analog that is widely used for RNA analysis, including short-range RNA-RNA crosslinking and nascent RNA labeling. The crosslinking thio moiety is attached directly to the nucleotide base, thus 4-SU Purity 99.89%
Catalog No. 50-287-8729 Supplier TARGETMOL CHEMICALS INC Supplier No. T3564825MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. 4-Thiouridine (4-SU) is a photoactivatable ribonucleoside analog that is widely used for RNA analysis, including short-range RNA-RNA crosslinking and nascent RNA labeling. The crosslinking thio moiety is attached directly to the nucleotide base, thus 4-SU Purity 99.89%