Learn More
Medchemexpress LLC 4-thiouridine 5′-triphosphate tetralithium | 20188-72-1 | 87.5% | 523.9 g/mol | C9H11Li4N2O14P3S | 10 MG
SDP
Catalog No. 5000287434
Encompass_Preferred
4-Thiouridine 5′-triphosphate tetralithium is a thiolated uridine triphosphate analogue used as a biochemical reagent for incorporation into RNA, cross-linking experiments, and transcriptional labeling. Supplied as the tetralithium salt, it is intended for research applications in molecular biology and nucleotide chemistry.
  • Used for nucleotide incorporation and transcriptional labeling.
  • Enables efficient cross-linking for RNA-protein interaction studies.
  • Provided as the tetralithium salt for improved solubility in aqueous buffers.
  • Available in small pack sizes suitable for laboratory research.
  • Characterized by CAS number 20188-72-1 and molecular formula C9H11Li4N2O14P3S.
Catalog No. 50-002-87434 Supplier Medchemexpress LLC Supplier No. HY15484010MG
4-Thiouridine 5′-triphosphate tetralithium is a thiolated uridine triphosphate analogue used as a biochemical reagent for incorporation into RNA, cross-linking experiments, and transcriptional labeling. Supplied as the tetralithium salt, it is intended for research applications in molecular biology and nucleotide chemistry.
  • Used for nucleotide incorporation and transcriptional labeling.
  • Enables efficient cross-linking for RNA-protein interaction studies.
  • Provided as the tetralithium salt for improved solubility in aqueous buffers.
  • Available in small pack sizes suitable for laboratory research.
  • Characterized by CAS number 20188-72-1 and molecular formula C9H11Li4N2O14P3S.